accessibility menu, dialog, popup

UserName

CAS RN 886497-73-0

CH3OH3CF

CAS RN 886497-73-0

IUPAC Name: 1-(3-fluoro-5-methylphenyl)ethan-1-one
Synonyms: 1-(3-fluoro-5-methylphenyl)ethanone
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: KFJMZDKKROJRHE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results