missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11345369
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345369 5 g 5g
11335369 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345369 Supplier Thermo Scientific Alfa Aesar Supplier No. H50383.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67475-55-2
Molecular Formula C7H6ClFO3S
Molecular Weight (g/mol) 224.63
MDL Number MFCD02091380
InChI Key QIVDOYCKCLEKPL-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzene-1-sulfonyl chloride, 3-fluoro-4-methoxy-benzenesulfonyl chloride, 3-fluoro-4-methoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methoxy, chloro 3-fluoro-4-methoxyphenyl sulfone, pubchem5161, 4-chlorosulphonyl-2-fluoroanisole, 3-fluoro-4-methoxyphenylsulfonyl chloride, 3-fluoro-4-methoxy-benzenesulphonyl chloride, 3-fluoranyl-4-methoxy-benzenesulfonyl chloride
PubChem CID 2774540
IUPAC Name 3-fluoro-4-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)F

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 224.64
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.