accessibility menu, dialog, popup

UserName

H3COSOOClF

CAS RN 67475-55-2

IUPAC Name: 3-fluoro-4-methoxybenzene-1-sulfonyl chloride
Synonyms: 3-fluoro-4-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 224.63
Molecular Formula: C7H6ClFO3S
InChi Key: QIVDOYCKCLEKPL-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results