missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylcinnamic acid, 97%, Thermo Scientific™

Catalog Number 15425527
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
155814-23-6
C10H9FO2
180.18
MFCD06205416
NZDLRLIWXWVPES-UHFFFAOYSA-N
2-propenoic acid, 3-3-fluoro-2-methylphenyl-, 2e, 3-3-fluoro-2-methylphenyl prop-2-enoic acid
54138108
3-(3-fluoro-2-methylphenyl)prop-2-enoic acid
CC1=C(C=CC(O)=O)C=CC=C1F
This item is not returnable. View return policy
155814-23-6
C10H9FO2
180.18
MFCD06205416
NZDLRLIWXWVPES-UHFFFAOYSA-N
2-propenoic acid, 3-3-fluoro-2-methylphenyl-, 2e, 3-3-fluoro-2-methylphenyl prop-2-enoic acid
54138108
3-(3-fluoro-2-methylphenyl)prop-2-enoic acid
CC1=C(C=CC(O)=O)C=CC=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 212°C to 215°C
Quantity 250 mg
Formula Weight 180.18
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-methylcinnamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'