missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl Haloperidol Decanoate, TRC

Product Code. 30382242
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382242 25 mg
30381336 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382242 Supplier Toronto Research Chemicals Supplier No. TRCE91871025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797982-02-5
Molecular Formula C33H45ClFNO3
Molecular Weight (g/mol) 558.17
IUPAC Name 4-(4-chlorophenyl)-1-(4-(3-ethyl-4-fluorophenyl)-4-oxobutyl)piperidin-4-yl decanoate
SMILES O=C(C1=CC(CC)=C(C=C1)F)CCCN2CCC(C3=CC=C(C=C3)Cl)(OC(CCCCCCCCC)=O)CC2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C33H45ClFNO3/c1-3-5-6-7-8-9-10-13-32(38)39-33(28-15-17-29(34)18-16-28)20-23-36(24-21-33)22-11-12-31(37)27-14-19-30(35)26(4-2)25-27/h14-19,25H,3-13,20-24H2,1-2H3
Formula Weight 557.31
Chemical Name or Material 3-Ethyl Haloperidol Decanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.