CAS RN 1797982-02-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Ethyl Haloperidol Decanoate, TRC
CAS: 1797982-02-5 Molecular Formula: C33H45ClFNO3 Molecular Weight (g/mol): 558.17 IUPAC Name: 4-(4-chlorophenyl)-1-(4-(3-ethyl-4-fluorophenyl)-4-oxobutyl)piperidin-4-yl decanoate SMILES: O=C(C1=CC(CC)=C(C=C1)F)CCCN2CCC(C3=CC=C(C=C3)Cl)(OC(CCCCCCCCC)=O)CC2