missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzhydrazide, 98%, Thermo Scientific™

Product Code. 11352466
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352466 1 g 1g
11362466 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352466 Supplier Thermo Scientific Alfa Aesar Supplier No. L06330.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Ethoxybenzhydrazide is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27830-16-6
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00017063
InChI Key ROKJUCOMVRXSAC-UHFFFAOYSA-N
Synonym 3-ethoxybenzhydrazide, m-ethoxy benzhydrazide, 3-ethoxybenzoic acid hydrazide, 1-ethoxybenzene-3-carbohydrazide, m-ethoxybenzhydrazide, #, acmc-1cs7h, 3-ethoxybenzenecarbohydrazide, benzoic acid,3-ethoxy-, hydrazide
PubChem CID 141381
IUPAC Name 3-ethoxybenzohydrazide
SMILES CCOC1=CC=CC(=C1)C(=O)NN

Specifications

Melting Point 101°C to 104°C
Quantity 1 g
Beilstein 2577763
Solubility Information Insoluble in water.
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material 3-Ethoxybenzhydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.