accessibility menu, dialog, popup

UserName

CH3OOHNH2N

CAS RN 27830-16-6

IUPAC Name: 3-ethoxybenzohydrazide
Synonyms: 3-ethoxybenzohydrazide
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: ROKJUCOMVRXSAC-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results