missing translation for 'onlineSavingsMsg'
Learn More

3’-Deoxyguanosine (>90%), TRC

Product Code. 30351200
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351200 25 mg 25mg
30359370 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351200 Supplier Toronto Research Chemicals Supplier No. TRCD24242025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3608-58-0
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.24
Synonym 9-(3-Deoxy-β-D-erythro-pentofuranosyl)-Guanine
IUPAC Name 2-amino-9-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one
SMILES OC[C@@H]1C[C@H]([C@H](N2C=NC3=C2N=C(N)NC3=O)O1)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13N5O4/c11-10-13-7-6(8(18)14-10)12-3-15(7)9-5(17)1-4(2-16)19-9/h3-5,9,16-17H,1-2H2,(H3,11,13,14,18)/t4-,5+,9+/m0/s1
Formula Weight 267.1
Chemical Name or Material 3’-Deoxyguanosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.