missing translation for 'onlineSavingsMsg'
Learn More

3-Deoxy-D-manno-2-octulosonic Acid Ammonium Salt, TRC

Product Code. 30358070
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358070 10 mg 10mg
30354591 50 mg 50mg
30356823 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358070 Supplier Toronto Research Chemicals Supplier No. TRCD24200010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-70-2
Molecular Formula C8 H14 O8 . H3 N
Molecular Weight (g/mol) 255.22
IUPAC Name azane;(4R,5R,6R,7R)-4,5,6,7,8-pentahydroxy-2-oxooctanoic acid
SMILES N.OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CC(=O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H14O8.H3N/c9-2-5(12)7(14)6(13)3(10)1-4(11)8(15)16;/h3,5-7,9-10,12-14H,1-2H2,(H,15,16);1H3/t3-,5-,6-,7-;/m1./s1
Formula Weight 255.0954
Percent Purity >95%
Chemical Name or Material 3-Deoxy-D-manno-2-octulosonic Acid, Ammonium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.