CAS RN 103404-70-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Deoxy-D-manno-2-octulosonic Acid Ammonium Salt, TRC
CAS: 103404-70-2 Molecular Formula: C8 H14 O8 . H3 N Molecular Weight (g/mol): 255.22 IUPAC Name: azane;(4R,5R,6R,7R)-4,5,6,7,8-pentahydroxy-2-oxooctanoic acid SMILES: N.OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CC(=O)C(=O)O