missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzoyl chloride, 98%

Product Code. 10107502 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10107502 1 g Glass bottle 1g
10682075 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107502 Supplier Thermo Scientific Acros Supplier No. 378130010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1711-11-1 | C8H4ClNO | 165.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-11-1
Molecular Formula C8H4ClNO
Molecular Weight (g/mol) 165.58
MDL Number MFCD00011545
InChI Key RPESZQVUWMFBEO-UHFFFAOYSA-N
Synonym 3-cyanobenzoylchloride, m-cyanobenzoyl chloride, unii-m7zy44hlss, m7zy44hlss, benzoyl chloride, 3-cyano, 3-cyanobenzoic acid chloride, 3-cyanobenzoyl-chloride, pubchem10466, acmc-20an5i, 3-cyano-benzoyl chloride
PubChem CID 308483
IUPAC Name 3-cyanobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 44°C
Color Gray or White to Yellow
Boiling Point 125°C to 129°C (11.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NCC6H4COCl
Quantity 1 g
Formula Weight 165.58
Percent Purity 98%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Chemical Name or Material 3-Cyanobenzoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Cyanobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.