accessibility menu, dialog, popup

UserName

ClON

CAS RN 1711-11-1

IUPAC Name: 3-cyanobenzoyl chloride
Synonyms: 3-cyanobenzoyl chloride
Molecular Weight (g/mol): 165.58
Molecular Formula: C8H4ClNO
InChi Key: RPESZQVUWMFBEO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(=C1)C#N
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)