missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenyl isothiocyanate, 97%

Product Code. 11434044
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434044 5 g 5g
11444044 25 g 25g
11454044 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434044 Supplier Thermo Scientific Alfa Aesar Supplier No. A11999.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2392-68-9 | C7H4ClNS | 169.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2392-68-9
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.63
MDL Number MFCD00004805
InChI Key WGXCKFMVBAOIFH-UHFFFAOYSA-N
Synonym 3-chlorophenyl isothiocyanate, 3-chlorophenylisothiocyanate, benzene, 1-chloro-3-isothiocyanato, isothiocyanic acid, 3-chlorophenyl ester, isothiocyanic acid, m-chlorophenyl ester, m-chlorophenyl isothiocyanate, 1-chloro-3-isothiocyanato-benzene, 3-chlorobenzenisothiocyanate, wln: scnr cg, acmc-1cfb8
PubChem CID 75449
IUPAC Name 1-chloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(=C1)N=C=S

Specifications

Density 1.281
Quantity 5 g
Odor Pungent
Refractive Index 1.6585
UN Number UN2922
Beilstein 636856
Boiling Point 249°C to 250°C
Formula Weight 169.64
Percent Purity 97%
Chemical Name or Material 3-Chlorophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.