CAS RN 2392-68-9
CAS RN 2392-68-9
IUPAC Name:
1-chloro-3-isothiocyanatobenzene
Synonyms:
1-chloro-3-isothiocyanatobenzene
Molecular Weight (g/mol):
169.63
Molecular Formula:
C7H4ClNS
InChi Key:
WGXCKFMVBAOIFH-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(=C1)N=C=S
1
–
1
of
1
results
1
3-Chlorophenyl isothiocyanate, 97%
CAS: 2392-68-9 Molecular Formula: C7H4ClNS Molecular Weight (g/mol): 169.63 MDL Number: MFCD00004805 InChI Key: WGXCKFMVBAOIFH-UHFFFAOYSA-N Synonym: 3-chlorophenyl isothiocyanate,3-chlorophenylisothiocyanate,benzene, 1-chloro-3-isothiocyanato,isothiocyanic acid, 3-chlorophenyl ester,isothiocyanic acid, m-chlorophenyl ester,m-chlorophenyl isothiocyanate,1-chloro-3-isothiocyanato-benzene,3-chlorobenzenisothiocyanate,wln: scnr cg,acmc-1cfb8 PubChem CID: 75449 IUPAC Name: 1-chloro-3-isothiocyanatobenzene SMILES: ClC1=CC=CC(=C1)N=C=S