missing translation for 'onlineSavingsMsg'
Learn More

3-chloroisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10351970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351970 5 g Glass bottle 5g
10411250 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351970 Supplier Thermo Scientific Acros Supplier No. 433750050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 88912-27-0
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD03094685
InChI Key MYAZXWFEMDJTFE-UHFFFAOYSA-N
Synonym 3-chloroisonicotinic acid, 3-chloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-chloro, 3-chloro-4-pyridinecarboxylicacid, pubchem5087, acmc-209qxw, 3-chloroisonicotinic cid, 3-chloro-isonicotinic acid, ksc447s3l
PubChem CID 2735816
IUPAC Name 3-chloropyridine-4-carboxylic acid
SMILES C1=CN=CC(=C1C(=O)O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 227°C to 229°C
Color Black to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 157.56
Percent Purity 97%
Physical Form Solution
Chemical Name or Material 3-Chloroisonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloroisonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.