CAS RN 88912-27-0
IUPAC Name:
3-chloropyridine-4-carboxylic acid
Synonyms:
3-chloropyridine-4-carboxylic acid
Molecular Weight (g/mol):
157.55
Molecular Formula:
C6H4ClNO2
InChi Key:
MYAZXWFEMDJTFE-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Cl)C=NC=C1
1
–
1
of
1
results
1
3-chloroisonicotinic acid, 97%, Thermo Scientific™
CAS: 88912-27-0 Molecular Formula: C6H4ClNO2 Molecular Weight (g/mol): 157.56 MDL Number: MFCD03094685 InChI Key: MYAZXWFEMDJTFE-UHFFFAOYSA-N Synonym: 3-chloroisonicotinic acid,3-chloro-4-pyridinecarboxylic acid,4-pyridinecarboxylic acid, 3-chloro,3-chloro-4-pyridinecarboxylicacid,pubchem5087,acmc-209qxw,3-chloroisonicotinic cid,3-chloro-isonicotinic acid,ksc447s3l PubChem CID: 2735816 IUPAC Name: 3-chloropyridine-4-carboxylic acid SMILES: C1=CN=CC(=C1C(=O)O)Cl