Learn More
3-Chloro-4-methylphenol, 97%, Thermo Scientific Chemicals
£63.30 - £185.00
Chemical Identifiers
CAS | 615-62-3 |
---|---|
Molecular Formula | C7H7ClO |
Molecular Weight (g/mol) | 142.58 |
MDL Number | MFCD00060319 |
InChI Key | VQZRLBWPEHFGCD-UHFFFAOYSA-N |
Synonym | phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol |
PubChem CID | 14853 |
IUPAC Name | 3-chloro-4-methylphenol |
SMILES | CC1=CC=C(O)C=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377179
|
Thermo Scientific Alfa Aesar
H27385.06 |
5 g |
£63.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387179
|
Thermo Scientific Alfa Aesar
H27385.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
615-62-3 | |
142.58 | |
VQZRLBWPEHFGCD-UHFFFAOYSA-N | |
14853 | |
CC1=CC=C(O)C=C1Cl |
C7H7ClO | |
MFCD00060319 | |
phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol | |
3-chloro-4-methylphenol |
Specifications
615-62-3 | |
228°C | |
MFCD00060319 | |
phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol | |
CC1=CC=C(O)C=C1Cl | |
142.58 | |
142.59 | |
3-Chloro-4-methylphenol |
51°C to 53°C | |
C7H7ClO | |
UN3437 | |
VQZRLBWPEHFGCD-UHFFFAOYSA-N | |
3-chloro-4-methylphenol | |
14853 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
missing translation for 'einecsNumber' : 210-439-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only