missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenol, 97%, Thermo Scientific Chemicals

Product Code. 11377179
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377179 5 g 5g
11387179 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377179 Supplier Thermo Scientific Alfa Aesar Supplier No. H27385.06
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-62-3 | Molecular Formula: C7H7ClO | Molecular Weight: 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-62-3
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00060319
InChI Key VQZRLBWPEHFGCD-UHFFFAOYSA-N
Synonym phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol
PubChem CID 14853
IUPAC Name 3-chloro-4-methylphenol
SMILES CC1=CC=C(O)C=C1Cl

Specifications

Melting Point 51°C to 53°C
Boiling Point 228°C
Quantity 5 g
UN Number UN3437
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.