missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylaniline hydrochloride, 97%, Thermo Scientific™

Product Code. 15494067
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494067 25 g 25g
15484067 5 g 5g
15404077 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494067 Supplier Thermo Scientific Alfa Aesar Supplier No. H31342.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7745-89-3
Molecular Formula C7H9Cl2N
Molecular Weight (g/mol) 178.056
MDL Number MFCD00242925
InChI Key YOQUZCNSUOJWQS-UHFFFAOYSA-N
Synonym 3-chloro-4-methylaniline hydrochloride, starlicide, caswell no. 216a, 3-chloro-p-toluidine hydrochloride, unii-7qq8474d5s, benzenamine, 3-chloro-4-methyl-, hydrochloride, epa pesticide chemical code 009901, p-toluidine, 3-chloro-, hydrochloride, 4-methyl, 3-chloroaniline hydrochloride, cth
PubChem CID 62651
IUPAC Name 3-chloro-4-methylaniline;hydrochloride
SMILES CC1=C(C=C(C=C1)N)Cl.Cl

Specifications

Melting Point ∼260°C (sublimation)
Quantity 25 g
Formula Weight 178.06
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-methylaniline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.