CAS RN 7745-89-3
IUPAC Name:
hydrogen 3-chloro-4-methylaniline chloride
Synonyms:
hydrogen 3-chloro-P-toluidine chloride
Molecular Weight (g/mol):
178.06
Molecular Formula:
C7H9Cl2N
InChi Key:
YOQUZCNSUOJWQS-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=CC=C(N)C=C1Cl
1
–
1
of
1
results
1
3-Chloro-4-methylaniline hydrochloride, 97%, Thermo Scientific™
CAS: 7745-89-3 Molecular Formula: C7H9Cl2N Molecular Weight (g/mol): 178.056 MDL Number: MFCD00242925 InChI Key: YOQUZCNSUOJWQS-UHFFFAOYSA-N Synonym: 3-chloro-4-methylaniline hydrochloride,starlicide,caswell no. 216a,3-chloro-p-toluidine hydrochloride,unii-7qq8474d5s,benzenamine, 3-chloro-4-methyl-, hydrochloride,epa pesticide chemical code 009901,p-toluidine, 3-chloro-, hydrochloride,4-methyl, 3-chloroaniline hydrochloride,cth PubChem CID: 62651 IUPAC Name: 3-chloro-4-methylaniline;hydrochloride SMILES: CC1=C(C=C(C=C1)N)Cl.Cl