missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-ethoxy-5-fluorobenzylamine, 97%, Thermo Scientific™

Product Code. 15404297
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404297 250 mg 250mg
15494287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404297 Supplier Thermo Scientific Alfa Aesar Supplier No. H31681.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-02-0
Molecular Formula C9H11ClFNO
Molecular Weight (g/mol) 203.641
MDL Number MFCD09832346
InChI Key QJCQLZGSGGNRRH-UHFFFAOYSA-N
Synonym 3-chloro-4-ethoxy-5-fluorobenzylamine, 1-3-chloro-4-ethoxy-5-fluorophenyl methanamine, 3-chloro-4-ethoxy-5-fluorobenzylamine, jrd, 3-chloro-4-ethoxy-5-fluorophenyl methanamine, 3-chloro-4-ethoxy-5-fluorophenyl methylamine
PubChem CID 46737662
IUPAC Name (3-chloro-4-ethoxy-5-fluorophenyl)methanamine
SMILES CCOC1=C(C=C(C=C1Cl)CN)F

Specifications

Refractive Index 1.5225
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 203.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-ethoxy-5-fluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.