accessibility menu, dialog, popup

UserName

CAS RN 1017779-02-0

CH3OClNH2F

CAS RN 1017779-02-0

IUPAC Name: 1-(3-chloro-4-ethoxy-5-fluorophenyl)methanamine
Synonyms: 1-(3-chloro-4-ethoxy-5-fluorophenyl)methanamine
Molecular Weight (g/mol): 203.64
Molecular Formula: C9H11ClFNO
InChi Key: QJCQLZGSGGNRRH-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(CN)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results