missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, TRC

Product Code. 30365250
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365250 50 mg 50mg
30363472 250 mg 250mg
30363211 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365250 Supplier Toronto Research Chemicals Supplier No. TRCC42856850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1029439-70-0
Molecular Formula C12H16BClO3
Molecular Weight (g/mol) 254.52
IUPAC Name 3-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES CC1(C)OB(OC1(C)C)c2ccc(O)cc2Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H16BClO3/c1-11(2)12(3,4)17-13(16-11)9-6-5-8(15)7-10(9)14/h5-7,15H,1-4H3
Formula Weight 254.0881
Chemical Name or Material 3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.