CAS RN 1029439-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, TRC
CAS: 1029439-70-0 Molecular Formula: C12H16BClO3 Molecular Weight (g/mol): 254.52 IUPAC Name: 3-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol SMILES: CC1(C)OB(OC1(C)C)c2ccc(O)cc2Cl