missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-2',6'-difluoroacetophenone, 97+%, Thermo Scientific™

Product Code. 11465897
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11465897 1 g 1g
11475897 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11465897

Brand: Thermo Scientific Alfa Aesar B24445.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an intermediate in organic syntheses.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177942-50-6
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD01631329
InChI Key GZGJXQFBPXHWRR-UHFFFAOYSA-N
Synonym 3'-chloro-2',6'-difluoroacetophenone, 1-3-chloro-2,6-difluorophenyl ethanone, 2',6'-difluoro-3'-chloroacetophenone, 3-chloro-2,6-difluoroacetophenone, 1-3-chloro-2,6-difluorophenyl ethan-1-one, 1-3-chloro-2,6-difluoro-phenyl-ethanone, 1-acetyl-3-chloro-2,6-difluorobenzene, 2,6-difluoro-3-chloroacetophenone, pubchem4317, pubchem7092
PubChem CID 2773505
IUPAC Name 1-(3-chloro-2,6-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1F)Cl)F

Specifications

Quantity 1 g
Formula Weight 190.58
Percent Purity ≥97%
Chemical Name or Material 3'-Chloro-2',6'-difluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.