accessibility menu, dialog, popup

UserName

CAS RN 177942-50-6

H3COFClF

CAS RN 177942-50-6

IUPAC Name: 1-(3-chloro-2,6-difluorophenyl)ethan-1-one
Synonyms: 1-(3-chloro-2,6-difluorophenyl)ethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: GZGJXQFBPXHWRR-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results