Learn More
3-Bromoquinoline, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 5332-24-1 |
---|---|
Molecular Formula | C9H6BrN |
Molecular Weight (g/mol) | 208.058 |
MDL Number | MFCD00006767 |
InChI Key | ZGIKWINFUGEQEO-UHFFFAOYSA-N |
Synonym | quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i |
PubChem CID | 21413 |
IUPAC Name | 3-bromoquinoline |
SMILES | C1=CC=C2C(=C1)C=C(C=N2)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11451968
|
Thermo Scientific Alfa Aesar
A14573.09 |
10 g |
N/A
|
N/A | |||||
11461968
|
Thermo Scientific Alfa Aesar
A14573.18 |
50 g |
N/A
|
N/A | |||||
11379509
|
Thermo Scientific Alfa Aesar
A14573.22 |
100 g |
N/A
|
N/A | |||||
11471968
|
Thermo Scientific Alfa Aesar
A14573.30 |
250 g |
N/A
|
N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5332-24-1 | |
208.058 | |
ZGIKWINFUGEQEO-UHFFFAOYSA-N | |
21413 | |
C1=CC=C2C(=C1)C=C(C=N2)Br |
C9H6BrN | |
MFCD00006767 | |
quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i | |
3-bromoquinoline |
Specifications
5332-24-1 | |
1.533 | |
>110°C (230°F) | |
1.664 | |
112939 | |
ZGIKWINFUGEQEO-UHFFFAOYSA-N | |
3-bromoquinoline | |
21413 | |
98% |
13°C to 15°C | |
274°C to 276°C | |
C9H6BrN | |
MFCD00006767 | |
quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i | |
C1=CC=C2C(=C1)C=C(C=N2)Br | |
208.058 | |
208.06 | |
3-Bromoquinoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 226-237-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only