missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoquinoline, 98%

Product Code. 11461968
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461968 50 g 50g
11451968 10 g 10g
11379509 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461968 Supplier Thermo Scientific Alfa Aesar Supplier No. A14573.18
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5332-24-1 | C9H6BrN | 208.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-24-1
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00006767
InChI Key ZGIKWINFUGEQEO-UHFFFAOYSA-N
Synonym quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
PubChem CID 21413
IUPAC Name 3-bromoquinoline
SMILES C1=CC=C2C(=C1)C=C(C=N2)Br

Specifications

Melting Point 13°C to 15°C
Density 1.533
Boiling Point 274°C to 276°C
Flash Point >110°C (230°F)
Refractive Index 1.664
Quantity 50 g
Beilstein 112939
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 3-Bromoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.