missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl alcohol, 99%

Product Code. 11414333
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414333 100 g 100g
11494323 5 g 5g
11404333 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414333 Supplier Thermo Scientific Alfa Aesar Supplier No. A10929.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 15852-73-0 | C7H7BrO | 187.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15852-73-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00004629
InChI Key FSWNRRSWFBXQCL-UHFFFAOYSA-N
Synonym 3-bromobenzyl alcohol, 3-bromophenyl methanol, m-bromobenzyl alcohol, benzenemethanol, 3-bromo, 3-bromobenzylalcohol, 3-bromophenyl methan-1-ol, 3-bromo-benzenemethanol, m-bromobenzylalcohol, 3-bromo benzylalcohol, pubchem3747
PubChem CID 85141
IUPAC Name (3-bromophenyl)methanol
SMILES C1=CC(=CC(=C1)Br)CO

Specifications

Density 1.56
Boiling Point 165°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.584
Quantity 100 g
Beilstein 2242512
Formula Weight 187.04
Percent Purity 99%
Chemical Name or Material 3-Bromobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.