CAS RN 15852-73-0
CAS RN 15852-73-0
IUPAC Name:
(3-bromophenyl)methanol
Synonyms:
(3-bromophenyl)methanol
Molecular Weight (g/mol):
187.04
Molecular Formula:
C7H7BrO
InChi Key:
FSWNRRSWFBXQCL-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC(Br)=C1
1
–
1
of
1
results
1
3-Bromobenzyl alcohol, 99%
CAS: 15852-73-0 Molecular Formula: C7H7BrO Molecular Weight (g/mol): 187.036 MDL Number: MFCD00004629 InChI Key: FSWNRRSWFBXQCL-UHFFFAOYSA-N Synonym: 3-bromobenzyl alcohol,3-bromophenyl methanol,m-bromobenzyl alcohol,benzenemethanol, 3-bromo,3-bromobenzylalcohol,3-bromophenyl methan-1-ol,3-bromo-benzenemethanol,m-bromobenzylalcohol,3-bromo benzylalcohol,pubchem3747 PubChem CID: 85141 IUPAC Name: (3-bromophenyl)methanol SMILES: C1=CC(=CC(=C1)Br)CO