missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxyphenylacetonitrile, 99%

Product Code. 11372287
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372287 2 g 2g
11382287 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372287 Supplier Thermo Scientific Alfa Aesar Supplier No. L09840.04
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 772-59-8 | C9H8BrNO | 226.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-59-8
Molecular Formula C9H8BrNO
Molecular Weight (g/mol) 226.07
MDL Number MFCD00016391
InChI Key OBJKHHRZMIIEOK-UHFFFAOYSA-N
PubChem CID 522655
IUPAC Name 2-(3-bromo-4-methoxyphenyl)acetonitrile
SMILES COC1=C(Br)C=C(CC#N)C=C1

Specifications

Melting Point 50°C to 53°C
Boiling Point 175°C to 178°C (6 mmHg)
Quantity 2 g
UN Number UN3439
Beilstein 2094194
Synonym 3-bromo-4-methoxyphenylacetonitrile, 3-bromo-4-methoxybenzyl cyanide, 2-3-bromo-4-methoxyphenyl acetonitrile, benzeneacetonitrile, 3-bromo-4-methoxy, 2-3-bromo-4-methoxyphenyl ethanenitrile, acmc-209p8t, benzeneacetonitrile,3-bromo-4-methoxy, 3-bromo-4-methoxy-phenyl-acetonitrile, 2-3-bromanyl-4-methoxy-phenyl ethanenitrile
Formula Weight 226.08
Percent Purity 99%
Chemical Name or Material 3-Bromo-4-methoxyphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.