CAS RN 772-59-8
IUPAC Name:
2-(3-bromo-4-methoxyphenyl)acetonitrile
Synonyms:
2-(3-bromo-4-methoxyphenyl)acetonitrile
Molecular Weight (g/mol):
226.07
Molecular Formula:
C9H8BrNO
InChi Key:
OBJKHHRZMIIEOK-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=C(CC#N)C=C1
1
–
1
of
1
results
1
3-Bromo-4-methoxyphenylacetonitrile, 99%
CAS: 772-59-8 Molecular Formula: C9H8BrNO Molecular Weight (g/mol): 226.07 MDL Number: MFCD00016391 InChI Key: OBJKHHRZMIIEOK-UHFFFAOYSA-N PubChem CID: 522655 IUPAC Name: 2-(3-bromo-4-methoxyphenyl)acetonitrile SMILES: COC1=C(Br)C=C(CC#N)C=C1