missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chloropentafluorobutyric acid, 97%, Thermo Scientific™

Product Code. 11399345
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399345 25 g 25g
11379345 1 g 1g
11389345 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399345 Supplier Thermo Scientific Alfa Aesar Supplier No. L16711.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216393-99-6
Molecular Formula C4HBrClF5O2
Molecular Weight (g/mol) 291.396
MDL Number MFCD00153071
InChI Key IFQBWLXWJWDSTE-UHFFFAOYSA-N
Synonym 3-bromo-4-chloropentafluorobutyric acid, acmc-1cmca, 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutyric acid, butanoic acid,3-bromo-4-chloro-2,2,3,4,4-pentafluoro, 3-bromanyl-4-chloranyl-2,2,3,4,4-pentakis fluoranyl butanoic acid
PubChem CID 2736232
IUPAC Name 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)Cl)(F)Br)(F)F)O

Specifications

Boiling Point 57°C (0.1 mmHg)
Quantity 25 g
UN Number UN3265
Formula Weight 291.4
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-chloropentafluorobutyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.