accessibility menu, dialog, popup

UserName

OFFClFBrFFHO

CAS RN 216393-99-6

IUPAC Name: 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutanoic acid
Synonyms: 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutanoic acid
Molecular Weight (g/mol): 291.4
Molecular Formula: C4HBrClF5O2
InChi Key: IFQBWLXWJWDSTE-UHFFFAOYNA-N
SMILES: OC(=O)C(F)(F)C(F)(Br)C(F)(F)Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results