missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzaldehyde, 97%

Product Code. 15471939
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471939 100 g 100g
15451939 5 g 5g
15461939 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471939 Supplier Thermo Scientific Alfa Aesar Supplier No. H64650.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149947-15-9 | C7H4BrFO | 203.01 g/mol

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149947-15-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD09027089
InChI Key OUAZPCKRSSEQKB-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorobenzenecarbaldehyde, 3-bromo-2-fluorobenzadehyde, 2-fluoro-3-bromobenzaldehyde, benzaldehyde,3-bromo-2-fluoro, benzaldehyde, 3-bromo-2-fluoro, 3-bromofluorobenzaldehyde, intermediates-zcf02145, acmc-209d3i, 3-bromo-2-fluoro-benzaldehyde, 3-bromanyl-2-fluoranyl-benzaldehyde
PubChem CID 22019361
IUPAC Name 3-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)F)C=O

Specifications

Melting Point 43°C to 46°C
Color White
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 13904499
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.