accessibility menu, dialog, popup

UserName

CAS RN 149947-15-9

IUPAC Name: 3-bromo-2-fluorobenzaldehyde
Synonyms: 3-bromo-2-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: OUAZPCKRSSEQKB-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results