missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzoyl chloride, 95%

Product Code. 11365419
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365419 1 g 1g
11355419 250 mg 250mg
15407877 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365419 Supplier Thermo Scientific Alfa Aesar Supplier No. H50410.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 61535-46-4 | C14H11ClO2 | 246.69 g/mol

Reactant for preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents, preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents, preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61535-46-4
Molecular Formula C14H11ClO2
Molecular Weight (g/mol) 246.69
MDL Number MFCD02258040
InChI Key HPVUGOBQRQUWMK-UHFFFAOYSA-N
Synonym 3-benzyloxy benzoyl chloride, 3-benzyloxybenzoyl chloride, 3-benzyloxy-benzoyl chloride, 3-benzyl-oxy benzoyl chloride, 3-phenylmethoxy benzoyl chloride, acmc-20ah7b, 3-benzyloxyl-benzoyl chloride, 3-benzyloxy-benzoic acid choride, benzoyl chloride, 3-phenylmethoxy
PubChem CID 4995375

Specifications

Melting Point 40°C to 42°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)C(=O)Cl
IUPAC Name 3-phenylmethoxybenzoyl chloride
Formula Weight 246.7
Percent Purity 95%
Chemical Name or Material 3-Benzyloxybenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.