accessibility menu, dialog, popup

UserName

OOCl

CAS RN 61535-46-4

IUPAC Name: 3-(benzyloxy)benzoyl chloride
Synonyms: 3-(benzyloxy)benzoyl chloride
Molecular Weight (g/mol): 246.69
Molecular Formula: C14H11ClO2
InChi Key: HPVUGOBQRQUWMK-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(OCC2=CC=CC=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results