missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid, 97%, Thermo Scientific™
£59.67 - £196.00
Chemical Identifiers
CAS | 2096333-24-1 |
---|---|
Molecular Formula | C16H17BFNO3 |
Molecular Weight (g/mol) | 301.12 |
MDL Number | MFCD20040268 |
InChI Key | XJQIDURCDMESBR-UHFFFAOYSA-N |
Synonym | 3-benzyl ethyl carbamoyl-4-fluorobenzeneboronic acid |
PubChem CID | 73996027 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2096333-24-1 | |
301.12 | |
XJQIDURCDMESBR-UHFFFAOYSA-N | |
73996027 |
C16H17BFNO3 | |
MFCD20040268 | |
3-benzyl ethyl carbamoyl-4-fluorobenzeneboronic acid |
Specifications
2096333-24-1 | |
MFCD20040268 | |
XJQIDURCDMESBR-UHFFFAOYSA-N | |
73996027 | |
97% |
C16H17BFNO3 | |
3-benzyl ethyl carbamoyl-4-fluorobenzeneboronic acid | |
301.12 | |
301.12 | |
3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid |
Sicherheit und Handhabung
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures