missing translation for 'onlineSavingsMsg'
Learn More

3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15401766
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2096333-24-1
C16H17BFNO3
301.12
MFCD20040268
XJQIDURCDMESBR-UHFFFAOYSA-N
3-benzyl ethyl carbamoyl-4-fluorobenzeneboronic acid
73996027
This item is not returnable. View return policy
2096333-24-1
C16H17BFNO3
301.12
MFCD20040268
XJQIDURCDMESBR-UHFFFAOYSA-N
3-benzyl ethyl carbamoyl-4-fluorobenzeneboronic acid
73996027

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 301.12
Percent Purity 97%
Chemical Name or Material 3-[Benzyl(ethyl)carbamoyl]-4-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'