missing translation for 'onlineSavingsMsg'
Learn More

(±)-3-Amino-iso-butyric-2,3,3-d3 Acid, TRC

Product Code. 30180430
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180430 10 mg 10mg
30179799 2 mg 2mg
30180097 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30180430 Supplier Toronto Research Chemicals Supplier No. TRCA60304410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219803-65-2
Molecular Formula C4 D3 H6 N O2
Molecular Weight (g/mol) 106.14
Synonym 3-Amino-2-methylpropanoic-2,3,3-d3 acid, 3-Amino-2-methylpropanoic acid-d3, 3-Amino-2-methylpropanoic acid 2,3,3-D3, 3-Amino-2-methylpropionic acid-d3, β-Aminoisobutyric acid-d3, 2-Methyl-β-alanine-2,3,3-d3
IUPAC Name 3-amino-2,3,3-trideuterio-2-methylpropanoic acid
SMILES [2H]C([2H])(N)C([2H])(C)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H9NO2/c1-3(2-5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/i2D2,3D
Formula Weight 106.0822
Percent Purity >95%
Chemical Name or Material (±)-3-Amino-iso-butyric-2,3,3-d3 Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.