CAS RN 1219803-65-2
CAS RN 1219803-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(±)-3-Amino-iso-butyric-2,3,3-d3 Acid, TRC
CAS: 1219803-65-2 Molecular Formula: C4 D3 H6 N O2 Molecular Weight (g/mol): 106.14 Synonym: 3-Amino-2-methylpropanoic-2,3,3-d3 acid,3-Amino-2-methylpropanoic acid-d3,3-Amino-2-methylpropanoic acid 2,3,3-D3,3-Amino-2-methylpropionic acid-d3,β-Aminoisobutyric acid-d3,2-Methyl-β-alanine-2,3,3-d3 IUPAC Name: 3-amino-2,3,3-trideuterio-2-methylpropanoic acid SMILES: [2H]C([2H])(N)C([2H])(C)C(=O)O
2
(±)-3-Amino-iso-butyric-2,3,3-d3 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.