missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(p-chlorophenyl)propionic acid, 98%, Thermo Scientific Chemicals

Product Code. p-55372 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10718851 1 g Glass Bottle 1g
10031821 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10718851 Supplier Thermo Scientific Acros Supplier No. 334330010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19947-39-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.64
MDL Number MFCD00126370
InChI Key BXGDBHAMTMMNTO-UHFFFAOYSA-N
Synonym 3-amino-3-4-chlorophenyl propanoic acid, 3-amino-3-4-chlorophenyl propionic acid, 3-p-chlorophenyl-dl-beta-alanine, 3-amino-3-4-chloro-phenyl-propionic acid, dl-beta-p-chlorophenyl alanine, 3-amino-3-p-chlorophenyl propionic acid, dl-3-amino-3-4-chlorophenyl-propionic acid, dl-beta-4-chlorophenyl alanine, rac-3-amino-3-4-chlorophenyl propionic acid, dl-3-amino-3-4'-chlorophenyl propionic acid
PubChem CID 344176
IUPAC Name 3-amino-3-(4-chlorophenyl)propanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)N)Cl

Specifications

Melting Point 243°C to 245°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula ClC6H4CH(NH2)CH2CO2H
Quantity 1 g
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Amino-3-(p-chlorophenyl)propionic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.