accessibility menu, dialog, popup

UserName

NH3(R)OOCl

CAS RN 19947-39-8

IUPAC Name: (3R)-3-azaniumyl-3-(4-chlorophenyl)propanoate
Synonyms: (3R)-3-ammonio-3-(4-chlorophenyl)propanoate
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: BXGDBHAMTMMNTO-MRVPVSSYSA-N
SMILES: [NH3+][C@H](CC([O-])=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)