missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(4-chlorophenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 11359765
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359765 250 mg 250mg
11339765 1 g 1g
11349765 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359765 Supplier Thermo Scientific Alfa Aesar Supplier No. L19798.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19947-39-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.634
MDL Number MFCD00126370
InChI Key BXGDBHAMTMMNTO-UHFFFAOYSA-N
Synonym 3-amino-3-4-chlorophenyl propanoic acid, 3-amino-3-4-chlorophenyl propionic acid, 3-p-chlorophenyl-dl-beta-alanine, 3-amino-3-4-chloro-phenyl-propionic acid, dl-beta-p-chlorophenyl alanine, 3-amino-3-p-chlorophenyl propionic acid, dl-3-amino-3-4-chlorophenyl-propionic acid, dl-beta-4-chlorophenyl alanine, rac-3-amino-3-4-chlorophenyl propionic acid, dl-3-amino-3-4'-chlorophenyl propionic acid
PubChem CID 344176
IUPAC Name 3-amino-3-(4-chlorophenyl)propanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)N)Cl

Specifications

Melting Point 214°C to 216°C (decomposition)
Beilstein 2365902
Quantity 250 mg
Formula Weight 199.64
Percent Purity 97%
Chemical Name or Material 3-Amino-3-(4-chlorophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.