missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-oxazolidinone, TRC

Product Code. 30136175
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136175 1 g 1g
17940815 100 mg 100mg
17934135 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136175 Supplier Toronto Research Chemicals Supplier No. TRCA6185701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-65-9
Molecular Formula C3 H6 N2 O2
Molecular Weight (g/mol) 102.09
Synonym 3-Amino-2-oxazolidinone, 3-Amino-1,3-oxazolidin-2-one, 3-Amino-2-oxazolidone, NSC 111187, NSC 196570, NSC 38250
IUPAC Name 3-amino-1,3-oxazolidin-2-one
SMILES NN1CCOC1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H6N2O2/c4-5-1-2-7-3(5)6/h1-2,4H2
Formula Weight 102.0429
Chemical Name or Material 3-Amino-2-oxazolidinone (AOZ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.