CAS RN 80-65-9
CAS RN 80-65-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Amino-2-oxazolidinone, TRC
CAS: 80-65-9 Molecular Formula: C3 H6 N2 O2 Molecular Weight (g/mol): 102.09 Synonym: 3-Amino-2-oxazolidinone,3-Amino-1,3-oxazolidin-2-one,3-Amino-2-oxazolidone,NSC 111187,NSC 196570,NSC 38250 IUPAC Name: 3-amino-1,3-oxazolidin-2-one SMILES: NN1CCOC1=O
2
AOZ, Fluka™
Analytical standard, reference material