missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Amino-2',3'-dideoxyguanosine, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3′-Amino-2′,3′-dideoxyguanosineis used as a pharmaceutical intermediate.
Solubility
Soluble in water (50 mg/ml).
Notes
Store in cool, dry place in a tightly sealed container. Store
3′-Amino-2′,3′-dideoxyguanosineis used as a pharmaceutical intermediate.
Solubility
Soluble in water (50 mg/ml).
Notes
Store in cool, dry place in a tightly sealed container. Store
Specifications
Specifications
| CAS | 66323-49-7 |
| Color | White to Yellow |
| Molecular Formula | C10H14N6O3 |
| MDL Number | MFCD09750824 |
| Quantity | 1 g |
| Synonym | 3'-amino-2',3'-dideoxyguanosine, 3'-nh2-ddg, guanosine, 3'-amino-2',3'-dideoxy, 2-amino-9-2r,4s,5s-4-amino-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 2-amino-9-2r,4s,5s-4-amino-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,4s,5s-4-amino-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, 2',3'-dideoxy-3'-aminoguanosine, 3'-amino-2'-deoxy-d-guanosine?, 3?-amino-2?,3?-dideoxyguanosine, 3'-amino-2', 3'-dideoxyguanosine |
| Solubility Information | Soluble in water (50mg/ml). |
| InChI Key | HOQBPAWPBCAUGA-DWFKHXNSNA-N |
| SMILES | N[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C1N=C(N)NC2=O |
| IUPAC Name | 2-amino-9-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction