missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

Product Code. 15403827
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15403827

Brand: Alfa Aesar™ H28525.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 185057-50-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD03001673
InChI Key FPXJNSKAXZNWMQ-UHFFFAOYSA-N
Synonym 3-amino-1-n-cbz-pyrrolidine, 1-cbz-3-aminopyrrolidine, 1-cbz-3-amino pyrrolidine, 1-benzyloxycarbonyl-3-aminopyrrolidine, 3-amino-pyrrolidine-1-carboxylic acid benzyl ester, 1-n-cbz-3-aminopyrrolidine, 3-amino-1-cbz-pyrrolidine, phenylmethyl 3-aminopyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-amino-, phenylmethyl ester, n-cbz-3-amino-pyrrolidine
PubChem CID 2756614
IUPAC Name benzyl 3-aminopyrrolidine-1-carboxylate
SMILES C1CN(CC1N)C(=O)OCC2=CC=CC=C2

Specifications

Refractive Index 1.5485
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 220.27
Chemical Name or Material (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.